missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLNK Mouse anti-Human, Clone: OTI4C9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808517S
This item is not returnable.
View return policy
Description
BLNK Monoclonal antibody specifically detects BLNK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| BLNK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WV28 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29760 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C9 | |
| Unconjugated | |
| BLNK | |
| AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa | |
| Human recombinant protein fragment corresponding to amino acids 1-292 of human BLNK produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction