Learn More
BLNK Mouse anti-Human, Unconjugated, Clone: 1A1A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66953-1-IG-20UL
This item is not returnable.
View return policy
Description
The BLNK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BLNK. This antibody recognizes human, pig antigen. The BLNK antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BLNK | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig |
| Western Blot, ELISA | |
| 1A1A1 | |
| Unconjugated | |
| 20 μL | |
| 29760 | |
| IgG1 |