Learn More
BLNK Mouse anti-Human, Unconjugated, Clone: 1A1A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66953-1-IG-20UL
 View more versions of this product
Product Code. 16849275
  • € 145.00 / 20µL
Description
The BLNK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BLNK. This antibody recognizes human, pig antigen. The BLNK antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BLNK
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig
Western Blot, ELISA
1A1A1
Unconjugated
20 μL
29760