missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLOC1S1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 474.00
Specifications
| Antigen | BLOC1S1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618781
|
Novus Biologicals
NBP2-92484-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640952
|
Novus Biologicals
NBP2-92484-0.1ml |
0.1 mL |
€ 474.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BLOC1S1 Polyclonal antibody specifically detects BLOC1S1 in Mouse samples. It is validated for Western BlotSpecifications
| BLOC1S1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 2647 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| biogenesis of lysosomal organelles complex-1, subunit 1, BLOC subunit 1, BLOC-1 subunit 1, BLOS1MICoA, FLJ39337, FLJ97089, GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1, GCN5 general control of amino-acid synthesis 5-like 1, GCN5 general control of amino-acid synthesis 5-like 1 (yeast), GCN5L1biogenesis of lysosome-related organelles complex 1 subunit 1, GCN5-like protein 1, MGC87455, MTA1-interacting coactivator, Protein RT14, RT14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-153 of human BLOC1S1 (NP_001478.2). MLSRLLKEHQAKQNERKELQEKRRREAITAATCLTEALVDHLNVGVAQAYMNQRKLDHEVKTLQVQAAQFAKQTGQWIGMVENFNQALKEIGDVENWARSIELDMRTIATALEYVYKGQLQSAPS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title