Learn More
Benchmark Scientific™ Block for Centrifuge Tubes
Product Code. 17954402
Change view
Click to view available options
No. of Places:
15
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places unitSize
17954402 15 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17954402 Supplier Benchmark Scientific™ Supplier No. H50005MT
This block holds 15 x 5 mL conical microtubes. Made of aluminum, it is covered with a protective shield. Fits Benchmark MultiTherm.
  • Accommodates 15 x 5 mL conical microtubes
  • Protective shield protects users from burns
  • Fits MultiTherm H and HC Models
Specifications
Autoclavable Not Autoclavable
Chamber Material Aluminum
Holds 15 x 5 mL Microtubes, Conical
Material Aluminum
No. of Places 15
Type Block