Learn More
Tenak™ Blood Bag Envelope
Shop All TENAK Products
Click to view available options
Capacity (Metric):
500 mL
750 mL
Unit Size:
Pack of 50
Description
- Envelope for blood bag
- 17 mm thick
- Arctice picture on one side
- Made of lacquered cardboard
Specifications
Specifications
| Material | Cardboard |
| Capacity (Metric) | 500 mL |
| Closure Color | White |
| Height (Metric) | 17 mm |
| Includes | Pack of 50; 135 x 235 mm (L x W) |
| Label Type | Text Box |