Learn More
Tenak™ Blood Bag Envelope
Product Code. 12467110 Shop All TENAK Products
Click to view available options
Capacity (Metric):
500 mL
750 mL
Unit Size:
Pack of 50
Product Code. 12467110
Brand: Tenak™ TE40112
Blood bag envelopes suitable for use with standard bloodbags.
  • Envelope for blood bag
  • 17 mm thick
  • Arctice picture on one side
  • Made of lacquered cardboard
Specifications
Material Cardboard
Capacity (Metric) 750 mL
Closure Color White
Height (Metric) 17 mm
Includes Pack of 50; 135 x 275 mm (L x W)
Label Type Text Box