missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blood Group A1, A2 Antigen Monoclonal Antibody (87-G)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17671
This item is not returnable.
View return policy
Description
MA1-7671 detects A1, A2 human blood protein from human samples. MA1-7671 has been successfully used in ELISA, agglutination and inhibition assays. The MA1-7671 immunogen is A1, A2 human blood protein.
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Specifications
| Blood Group A1, A2 Antigen | |
| Monoclonal | |
| 1.0 mg/mL | |
| Antibody with 1.25 mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| Agglutination, ELISA, Inhibition Assays | |
| 87-G | |
| Unconjugated | |
| Blood Group A | |
| A1, A2 human blood protein. | |
| 100 μg | |
| Primary | |
| 4°C short term, -80°C long term | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction