missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Blood Group B Antigen Monoclonal Antibody (89-F)
GREENER_CHOICE

Product Code. 11575362
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11575362 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11575362 Supplier Invitrogen™ Supplier No. MA17672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-7672 detects B human blood group protein from human samples. MA1-7672 has been successfully used in ELISA, inhibition assays, and agglutination applications. The MA1-7672 immunogen is B human blood group protein. Store short-term at 4°C, long-term at -80°C.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group B Antigen
Applications Agglutination, ELISA, Inhibition Assays
Classification Monoclonal
Clone 89-F
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.3M NaCl, 1.25mM EDTA and 0.01% sodium azide; pH 7.2
Gene ABO
Gene Accession No. P16442
Gene Symbols ABO
Host Species Mouse
Immunogen B human blood group protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.