missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Blood Group B Antigen Monoclonal Antibody (Z5H-2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA17666

Product Code. 11515362

  • € 530.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-7666 detects B human blood protein from human samples. MA1-7666 has been successfully used in agglutination applications. The MA1-7666 immunogen is B human blood protein. Store short-term at 4ºC, long-term at -80ºC.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blood Group B Antigen
Monoclonal
1 mg/mL
Antibody with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2
P16442
Mouse
Protein A
RUO
28
4°C short term, -80°C long term
Liquid
Agglutination
Z5H-2
Unconjugated
ABO
ABO
B human blood protein.
100 μg
Primary
Human
Antibody
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.