missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blooms Syndrome Protein Blm Antibody, Alexa Fluor™ 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-324AF350
This item is not returnable.
View return policy
Description
Blooms Syndrome Protein Blm Polyclonal antibody specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for Western Blot
Specifications
| Blooms Syndrome Protein Blm | |
| Polyclonal | |
| Western Blot | |
| Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2 | |
| The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Bloom Syndrome using the numbering given in entry NP_000048.1 (GeneID 641). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| 641 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction