missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLTR-1 (Leukotriene B4 Receptor) Mouse anti-Human, BUV615, Clone: 203/14F11 (also known as 14F11), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751224
This item is not returnable.
View return policy
Description
BLTR-1 (Leukotriene B4 Receptor) Monoclonal antibody specifically detects BLTR-1 (Leukotriene B4 Receptor) in Human samples. It is validated for Flow Cytometry
Specifications
| BLTR-1 (Leukotriene B4 Receptor) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| 203/14F11 (also known as 14F11) | |
| Brilliant Ultraviolet 615 | |
| hBLTR-1; BLT1; BLTR; P2Y7; GPR16; LTBR1; P2RY7; CMKRL1; LTB4R1 | |
| Human BLTR-1-transfected cell line | |
| RUO | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction