Learn More
ABEBA™ Blue Autoclavable Clogs 9610
Product Code. 15166946
Click to view available options
Size:
41/42
43/44
Unit Size:
1 pair
Product Code. 15166946
Brand: ABEBA™ 961041/42
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Blue
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Blue
Size 41/42
Resistant To Static
Product Type Controlled Environments Shoes