Learn More
Spectrum™ Labs Blue Polypropylene Standard Closures
Product Code. 11425979
Click to view available options
Width (Metric):
23 mm
75 mm
Unit Size:
Pack of 10
Product Code. 11425979
Brand: Spectrum™ Labs 142935
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These polypropylene closures lock over the tubing, forming a sack ensuring a leak-proof seal
These polypropylene closures lock over the tubing, forming a sack ensuring a leak-proof seal. Regular closures will float in water. They eliminate the need for tying knots in dialysis tubing, a practice which can adversely affect the porosity of a membrane. Lighter than water, regular closures are normally used with weighted or magnetic weighted closures. Attach a regular closure to the top of the dialysis tubing and a weighted or magnetic weighted closure to the bottom for vertical floating. Not for use with cellulose ester membrane tubing. Autoclavable.
Specifications
Product Type Standard Closure
Material Polypropylene
Width (Metric) 23 mm
Autoclavable Autoclavable
Closure Material Polypropylene
Closure Type Standard Closure
Color Blue
Sterilization Method Autoclavable
Temperature Range (Metric) 0°C to 121°C
Quantity 10 Pk.
For Use With (Application) For standard grade RC dialysis tubing