Learn More
Kimberly-Clark™ Scott™ Medical Couch Covers
Protects medical couches and maintains hygiene in a healthcare environment. Kimberly-Clark™ Scott™ Medical Couch Covers can be used with a dispensing system to help control use and reduce wastage .
Brand: Kimberly-Clark™ 6023
Additional Details : Weight : 0.50000kg
Description
- 2-ply couch cover
- Strong, tear resistant and durable
- Ideal for protecting couches and maintaining high levels of hygiene in the healthcare environment
- Perforated
- Can be used with a SCOTT™ Couch Cover Wiper Dispenser to help control use and reduce wastage
Specifications
Clinical Rolls | |
ISO 14001, ISO 9001 | |
Roll | |
Kimberly-Clark Professional Rolled Hand Towel Dispenser | |
37 cm | |
6 rolls X 135 sheets | |
6.864 kg |
Blue | |
37 cm x 50 | |
For protecting couches and maintaining high levels of hygiene in the healthcare environment. | |
14.976 in. | |
2-ply | |
15.1 lbs. | |
50 cm |