missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLZF1 Monoclonal Antibody (2E10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520435
This item is not returnable.
View return policy
Description
Peptide Sequence: KLAKAVNSHL LGNVGINNQK KIPSTVEFCS TPAEKMAETV LRILDPVTCK ESSPDNPFFE SSPTTLLATK KNIGRFHPYT RYENITFNCC NHCRGELIAL.
Required for normal Golgi structure and for protein transport from the endoplasmic reticulum (ER) through the Golgi apparatus to the cell surface.
Specifications
| BLZF1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9H2G9 | |
| BLZF1 | |
| BLZF1 (NP_003657, 301 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2E10 | |
| Unconjugated | |
| BLZF1 | |
| 1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8548 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction