missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMAL1 Antibody (JM17-34), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-67191
This item is not returnable.
View return policy
Description
BMAL1 Monoclonal specifically detects BMAL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| BMAL1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
| Rabbit | |
| Protein A purified | |
| Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision | |
| 406 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| JM17-34 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| ARNTL | |
| Synthetic peptide within Human BMAL1 aa 550-599/ 626. (SwissProt: O00327 Human; SwissProt: Q9WTL8 Mouse; SwissProt: Q9EPW1 Rat) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction