missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMAL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122196
This item is not returnable.
View return policy
Description
BMAL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| BMAL1 | |
| Polyclonal | |
| Unconjugated | |
| Arntl | |
| Arnt3; ARNTL; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; basic-helix-loop-helix-PAS protein MOP3; bHLHe5; bHLH-PAS protein JAP3; BMAL1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; Brain and muscle ARNT-like 1; brain and muscle Arnt-like protein 1; CG8727 PA; class E basic helix-loop-helix protein 5; cycle; JAP3; LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1; member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; tic | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 29657 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9EPW1 | |
| Arntl | |
| Recombinant protein Aryl Hydrocarbon Receptor Nuclear Translocator Like Protein. The antigen corresponds to amino acid range 1-169 of the target protein. | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction