missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMERB1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97384MFV610
This item is not returnable.
View return policy
Description
BMERB1 Polyclonal antibody specifically detects BMERB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BMERB1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| chromosome 16 open reading frame 45, FLJ32618, hypothetical protein LOC89927 | |
| Produced in rabbits immunized with E. coli-derived Human BMERB1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 89927 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction