missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BMERB1 Polyclonal antibody specifically detects BMERB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | BMERB1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | chromosome 16 open reading frame 45, FLJ32618, hypothetical protein LOC89927 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human BMERB1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?