missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody (17A9), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80091AF532
This item is not returnable.
View return policy
Description
Bmf Monoclonal antibody specifically detects Bmf in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Bmf | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Recombinant full length mouse Bmf and synthetic peptides corresponding to aa 13-27 and aa 112-126 of mouse Bmf. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| 17A9 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction