missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody (17A9), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Bio-Techne NBP2-80091PE
This item is not returnable.
View return policy
Description
Bmf Monoclonal antibody specifically detects Bmf in Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Bmf | |
| Monoclonal | |
| PE | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Recombinant full length mouse Bmf and synthetic peptides corresponding to aa 13-27 and aa 112-126 of mouse Bmf. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 17A9 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction