missing translation for 'onlineSavingsMsg'
Learn More

Bmf Antibody (9G10), CoraFluor™ 1, Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-80090CL1

Product Code. 30155106

  • € 1020.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Bmf Monoclonal antibody specifically detects Bmf in Human,Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bmf
Monoclonal
CoraFluor 1
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065
Recombinant mouse Bmf.
0.1 mL
Apoptosis, Cancer, Tumor Suppressors
90427
Store at 4°C in the dark. Do not freeze.
IgG2a κ
Flow Cytometry, Immunoprecipitation, Western Blot
9G10
PBS
Rat
Protein G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.