missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76658AF405
This item is not returnable.
View return policy
Description
Bmf Polyclonal antibody specifically detects Bmf in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Bmf | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 405 | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Antibody was raised with a synthetic peptide corresponding to 15 amino acids near the amino terminus of human Bmf. The immunogen is located within the first 50 amino acids of Bmf. Amino Acid Squence: EPSQCVEELEDDV | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction