missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97262AF594
This item is not returnable.
View return policy
Description
Bmf Polyclonal specifically detects Bmf in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Bmf | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Produced in rabbits immunized with E. coli-derived Human Bmf fragment. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction