missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99460CL1
This item is not returnable.
View return policy
Description
Bmf Polyclonal antibody specifically detects Bmf in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Bmf | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Bmf. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction