missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody (OTI6F3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72311AF750
This item is not returnable.
View return policy
Description
Bmf Monoclonal specifically detects Bmf in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Bmf | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BMF | |
| Full length recombinant protein of human Bmf ( NP_001003940) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI6F3 | |
| Western Blot, Immunohistochemistry | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction