Learn More
BMH-21, MedChemExpress
Product Code. 16485886
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16485886
Brand: Medchem Express HY12484
MedChemExpress BMH-21 is a first-in-class DNA intercalator which inhibits RNA polymerase I (Pol I) transcription. BMH-21 possesses anticancer activity.
BMH-21 is a first-in-class DNA intercalator which inhibits RNA polymerase I (Pol I) transcription. BMH-21 possesses anticancer activity.
Specifications
Chemical Name or Material BMH-21
Physical Form Solid
CAS 896705-16-1
Color Orange
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H20N4O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 1 mg/mL (2.77 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN2C1=NC3=C(C=C4C(C=CC=C4)=C3)C2=O)NCCN(C)C
Molecular Weight (g/mol) 360.41
Formula Weight 360.41
Percent Purity 98.61%
Grade Research
Show More Show Less
Research purposes only