missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMI-1 Monoclonal Antibody (B3-G5)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA542412

Product Code. 17799023

  • € 518.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: Hela, HepG2, Jurkat, 293T, MCF-7, PC12, K562, NIH/3T3, A549, LOVO, human tonsil tissue, human colon cancer tissue, human breast cancer tissue, mouse kidney tissue, human kidney tissue, human brain tissue. Subcellular Location: Nucleus, Cytoplasm.

Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BMI-1
Monoclonal
2 mg/mL
PBS with 40% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
P25916, P35226
BMI1
Recombinant protein within human Bmi1 full sequence.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
B3-G5
Unconjugated
BMI1
AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1
Mouse
Protein A
RUO
12151, 307151, 648
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.