missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-15/GDF-9B Antibody (28A), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61935IR
This item is not returnable.
View return policy
Description
BMP-15/GDF-9B Monoclonal specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA.
Specifications
| BMP-15/GDF-9B | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BMP15 | |
| Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human BMP-15/GDF-9B | |
| 0.1 mL | |
| 9210 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 28A | |
| Western Blot, ELISA | |
| BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction