missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-15/GDF-9B Antibody (BMP15/4263), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08876AF647
This item is not returnable.
View return policy
Description
BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA
Specifications
| BMP-15/GDF-9B | |
| Monoclonal | |
| Alexa Fluor 647 | |
| BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human BMP-15/GDF-9B (Uniprot: O95972) | |
| 100 μg | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology, Signal Transduction, Stem Cells | |
| 9210 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| BMP15/4263 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction