missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-15 Recombinant Rabbit Monoclonal Antibody (6K5C3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535450
This item is not returnable.
View return policy
Description
Immunogen sequence: LHPFQISFRQ LGWDHWIIAP PFYTPNYCKG TCLRVLRDGL NSPNHAIIQN LINQLVDQSV PRPSCVPYKY VPISVLMIEA NGSILYKEYE GMIAESCTCR.
BMP15, or GDF9B is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily that are involved in embryonic development and adult tissue homeostasis. BMP15 is expressed exclusively in the oocyte. It is an oocyte-specific growth/differentiation factor that may be involved in oocyte maturation and follicular development. Defects in BMP15 are the cause of ovarian dysgenesis type 2. BMP15 could be used as an oogenesis marker to track human amniotic fluid stem cells differentiation into the oocyte-like cells.
Specifications
| BMP-15 | |
| Recombinant Monoclonal | |
| 0.31 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95972, Q9Z0L4 | |
| BMP15 | |
| A synthetic peptide corresponding to a sequence within amino acids 293-392 of human BMP15 (O95972). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6K5C3 | |
| Unconjugated | |
| BMP15 | |
| AU015375; AU018861; AU021453; BMP; BMP15; Bmp-15; Bone morphogenetic protein; Bone morphogenetic protein 15; C86824; C87336; GDF9B; GDF-9B; growth differentiation factor-9B; growth/differentiation factor 9B; ODG2; POF4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12155, 59302, 9210 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction