missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-2 Antibody (15B3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42651AF750
This item is not returnable.
View return policy
Description
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BMP-2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BMP2 | |
| Recombinant human BMP2 (283-396aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 15B3 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
| Mouse | |
| Protein A purified | |
| Biologically Active Proteins, Cytokine Research, Embryonic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 650.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction