missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-2/BMP-4 Monoclonal Antibody (100230)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA523764

Product Code. 15573452

  • € 588.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody shows 100% cross-reactivity with recombinant zebra fish (rzf) BMP-2 and rhBMP-4, 20%-40% cross-reactivity with rhBMP-3, -3b, -6, and no cross-reactivity with rhBMP-5, -10, or -15. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method. Western blot: 1 μg/mL, under non--reducing conditions only Neutralization: Measured by its ability to neutralize BMP¬2-induced alkaline phosphatase production in the ATDC5 mouse chondrogenic cell line. The Neutralization Dose (ND50) is typically 5-15 μg/mL in the presence of 1 μg/mL Recombinant Human BMP-2 and 2 μg/mL heparin. This antibody also neutralizes Human BMP-4 induced activity in the ATDC5 mouse chondrogenic cell line. The ND50 is approximately 3 μg/mL in the presence of 75 ng/mL of Recombinant Human BMP-4.

Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BMP-2/BMP-4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P12643, P12644
BMP2, BMP4
CHO-derived recombinant human BMP-2 Gln283-Arg396.
500 μg
Primary
Human
Antibody
IgG1
Neutralization, Western Blot
100230
Unconjugated
BMP2
2610024H22Rik; AI467020; AL117858; AW546137; BB189135; BDA2; BMP; BMP2; BMP-2; BMP2A; BMP-2A; BMPRII; BMPR-II; Bone morphogenetic protein; Bone morphogenetic protein 2; bone morphogenetic protein 2 precursor (BMP-2) (BMP-2A); bone morphogenetic protein 2A; H-BMP-2; morphogen
Mouse
Protein A/G
RUO
650, 652
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.