missing translation for 'onlineSavingsMsg'
Learn More

BMP-3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15508482
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508482 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508482 Supplier Invitrogen Supplier No. PA547986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BMP-3 Polyclonal Antibody

BMP-3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene BMP3
Gene Accession No. P12645
Gene Alias 9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3A; bone morphogenetic protein-3; osteogenin; PBMP3
Gene Symbols BMP3
Host Species Goat
Immunogen E. coli-derived recombinant human BMP-3 Gln363-Arg472.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 651
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.