missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597507
This item is not returnable.
View return policy
Description
BMP-3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The BMPs belong to the TGF-Beta superfamilyBMPs stimulate the production of bone matrix proteins and thereby alter stromal cell and osteoclast proliferation, a key step in bone marrow development. In addition to promoting ectopic bone and cartilage development, BMPs regulate the growth, differentiation, chemotaxis, proliferation, and apoptosis of various cell types, including mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells. BMPs also play a role in dorsal/ventral patterning. BMP3, highly expressed in lung, ovary and small intestine, participates in cartilage and bone formation. BMP3 and BMP2 genes map to conserved regions between human and mouse.
Specifications
| BMP-3 | |
| Polyclonal | |
| Unconjugated | |
| BMP3 | |
| 9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3A; bone morphogenetic protein-3; osteogenin; PBMP3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 651 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P12645 | |
| BMP3 | |
| Recombinant Human Bone morphogenetic protein 3 protein (362-472AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction