Learn More
Invitrogen™ BMP-5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104248
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BMP5.
TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
Specifications
| BMP-5 | |
| Polyclonal | |
| Unconjugated | |
| Bmp5 | |
| AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12160, 653 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22003, P49003 | |
| Bmp5 | |
| A synthesized peptide derived from human BMP5(Accession P22003), corresponding to amino acid residues A143-V193. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |