Learn More
PeproTech BMP-7 Monoclonal Antibody, PeproTech®
Mouse Monoclonal Antibody
Brand:  PeproTech 500-M123-500UG
Product Code. 17845287
  • € 178.00 / 500µg
Description
AA Sequence of recombinant protein: MANVAENSSS DQRQACKKHE LYVSFRDLGW QDWIIAPEGY AAYYCEGECA FPLNSYMNAT NHAIVQTLVH FINPETVPKP CCAPTQLNAI SVLYFDDSSN VILKKYRNMV VRACGCH. Preparation: Produced in mice immunized with highly pure Recombinant Human BMP-7. Anti-Human BMP-7-specific antibody was purified from cell culture by Protein G affinity chromatography.
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BMP-7
Monoclonal
PBS with no preservative
P18075
Bmp7
CHO cells-derived Recombinant Human BMP-7.
500 μg
Primary
Human
Antibody
ELISA
Unconjugated
Bmp7
BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
Mouse
Protein G
RUO
655
-20°C
Lyophilized