missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-7 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143197
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize precursor only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
Specifications
| BMP-7 | |
| Polyclonal | |
| Unconjugated | |
| Bmp7 | |
| BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 492315 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| 0 | |
| Bmp7 | |
| Peptide with sequence C-SLQDSHFLTDAD (aa 120-131). | |
| 100 μg | |
| Primary | |
| Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction