missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-8B Monoclonal Antibody (GT1017)
GREENER_CHOICE

Product Code. 16332244
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16332244 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16332244 Supplier Invitrogen™ Supplier No. MA527733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: human BMP8B-transfected 293T cells Predicted reactivity: Human (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BMP-8B encodes a secreted ligand of the TGF-beta (transforming growth factor-beta) superfamily of proteins. Ligands of this family bind various TGF-beta receptors leading to recruitment and activation of SMAD family transcription factors that regulate gene expression. The encoded preproprotein is proteolytically processed to generate each subunit of the disulfide-linked homodimer. The encoded protein stimulates thermogenesis in brown adipose tissue. Expression of this gene may be downregulated in pancreatic cancer. BMP-8B may have arose from a gene duplication event and its gene duplicate is also present on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-8B
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT1017
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Bmp8b
Gene Accession No. P34820
Gene Alias BMP; BMP8; BMP-8; Bmp8b; BMP-8B; Bone morphogenetic protein; bone morphogenetic protein 8 (osteogenic protein 2); bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); MGC131757; OP2; OP-2; Op3; Osteogenic protein 2
Gene Symbols Bmp8b
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human BMP8b. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.