missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-9 Antibody (16S11), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22034R
This item is not returnable.
View return policy
Description
BMP-9 Monoclonal specifically detects BMP-9 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| BMP-9 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GDF2 | |
| recombinant protein of the mature human BMP-9 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 16S11 | |
| Western Blot, ELISA | |
| BMP9BMP-9Bone morphogenetic protein 9, GDF-2, growth differentiation factor 2, growth/differentiation factor 2 | |
| Mouse | |
| Protein G purified | |
| Cytokine Research | |
| 2658 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction