Learn More
BMP5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13253-1-AP-150UL
 View more versions of this product
Product Code. 16851374
  • € 410.00 / 150µL
Description
This gene encodes a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. These proteins are synthesized as prepropeptides, cleaved, and then processed into dimeric proteins. This protein may act as an important signaling molecule within the trabecular meshwork and optic nerve head, and may play a potential role in glaucoma pathogenesis. This gene is differentially regulated during the formation of various tumors.
Specifications
Specifications
BMP5
Polyclonal
Unconjugated
BMP5
BMP 5, BMP5, bone morphogenetic protein 5
Rabbit
Antigen Affinity Chromatography
RUO
12160, 653
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22003, P49003
BMP5
BMP5 Fusion Protein Ag3952
150 μL
Primary
Human, Mouse