missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP6 Monoclonal Antibody (D2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551039
This item is not returnable.
View return policy
Description
BMP6 is a member of bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, Bmp6 has a proposed role in early development. In addition, the fact that BMP6 is closely related to BMP5 and BMP7 has lead to speculation of possible bone inductive activity.
Specifications
| BMP6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P22004 | |
| BMP6 | |
| Recombinant Human BMP6 protein, Ser375-His513 (Accession #P22004). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| D2 | |
| Unconjugated | |
| BMP6 | |
| BMP; BMP 6; BMP6; BMP-6; Bone morphogenetic protein; Bone morphogenetic protein 6; D13Wsu115e; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; Vgr; Vgr1; VGR-1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100155536, 654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction