missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMPER Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122195
This item is not returnable.
View return policy
Description
BMPER Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a secreted protein that interacts with, and inhibits bone morphogenetic protein (BMP) function. It has been shown to inhibit BMP2- and BMP4-dependent osteoblast differentiation and BMP-dependent differentiation of the chondrogenic cells. Mutations in this gene are associated with a lethal skeletal disorder, diaphanospondylodysostosis.
Specifications
| BMPER | |
| Polyclonal | |
| Unconjugated | |
| Bmper | |
| BMP-Binding Endothelial Regulator Protein; CRIM3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 73230 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8CJ69 | |
| Bmper | |
| Recombinant protein BMP Binding Endothelial Regulator. The antigen corresponds to amino acid range 40-298 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction