missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR-IA/ALK-3 Antibody (MM0053-8L7), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
BMPR-IA/ALK-3 Monoclonal antibody specifically detects BMPR-IA/ALK-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Immunogen | Human recombinant BMPR-IA/ALK-3 (extracellular domain) |
| Purification Method | Protein G purified |
| Quantity | 0.1 mL |
| Research Discipline | Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers |
| Primary or Secondary | Primary |
| Isotype | IgG2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?