missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 564929

Description
BMS 564929 is a highly potent and selective androgen receptor (AR) agonist (EC50 = 0.44 nM). Exhibits selectivity for ARs over estrogen, glucocorticoid and mineralocorticoid receptors and no significant binding to steroid hormone binding globulin or aromatase. Also exhibits selectivity for muscle over prostate ARs. Active in vivo and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-3-methyl-4-[(7R,7aS)-tetrahydro-7-hydroxy-1,3-dioxo-1H-pyrrolo[1,2-c]imidazol-2(3H)-yl]benzonitrile |
| CAS | 627530-84-1 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C14H12ClN3O3 |
| Formula Weight | Observed MW: 305.72 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?