missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537215
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 146 kDa. Purity is >95% by SDS-PAGE.
BMS1 (Ribosome biogenesis protein BMS1 homolog) is a 1,282 amino acid protein encoded by the human gene BMS1. BMS1 is a nuclear protein that belongs to the BMS1/TSR1 family (BMS1 subfamily). BMS1 is believed to act as a molecular switch during maturation of the 40S ribosomal subunit in the nucleolus. The 40S ribosomal subunit is an important member of the 80S ribosome complex, which also includes initiator tRNA and a 60S ribosomal subunit. The 80S ribosome is assembled by eukaryotic initiation factors (eIFs) at the initiation codon of mRNA in order to begin translation initiation. The joining of these ribosomal subunits requires eIF5B.
Specifications
| BMS1 | |
| Polyclonal | |
| Unconjugated | |
| Bms1 | |
| AA408648; ACC; AU020092; AU043373; BB007109; BC030906; BMS1; BMS1 homolog, ribosome assembly protein; BMS1 homolog, ribosome assembly protein (yeast); BMS1 ribosome biogenesis factor; BMS1, ribosome biogenesis factor; Bms1l; BMS1-like, ribosome assembly protein; KIAA0187; mKIAA0187; ribosome assembly protein BMS1 homolog; Ribosome biogenesis protein BMS1 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 213895, 362426, 9790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14692 | |
| Bms1 | |
| A synthetic peptide corresponding to residues in human BMS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction