Learn More
Eurofins DiscoverX Bmx Protein, Active
Description
BMX (also called ETK, Bone marrow tyrosine kinase gene in chromosome X protein, Epithelial and endothelial tyrosine kinase) is one of the non-receptor tyrosine kinases, located intracellularly. It is a src homolog and contains an SH3 and SH2 domain upstream from the tyrosine kinase domain. Unlike other members of the BTK family kinases, which are mostly hemopoietic cell-specific, BMX is preferentially expressed in epithelial and endothelial cells of large arteries. Inhibitors of BMX may be useful in the treatment of atherosclerosis and cancer.
GenBank NM_001721
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 79 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥63% |
| Protein | Bmx, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.