missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109246
This item is not returnable.
View return policy
Description
Positive Control: Rat liver tissue lysate, HepG2, LOVO, human liver tissue, human placenta tissue.
NIP1 (BNIP1) is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four products of unknown function. Transcript variant BNIP1 contains the entire coding region of the gene. This variant contains a fully conserved BH3 domain, which has been associated with pro-apoptotic function.
Specifications
| BNIP1 | |
| Polyclonal | |
| Unconjugated | |
| BNIP1 | |
| 2010005M06Rik; 5930429G21Rik; BCL2 interacting protein 1; BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1; BCL2/adenovirus E1B interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1-like 1; bnip1; bnip1 {ECO:0000312; Bnip1l1; EMBL:AAL50991.1, ECO:0000312; NIP1; RGD:620799}; Sec20; Sec20l; SNARE protein SEC20; transformation-related gene 8 protein; TRG8; TRG-8; Unknown (protein for MGC:133975); vesicle transport protein SEC20 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 662 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q12981 | |
| BNIP1 | |
| Recombinant protein within Human BN1P1 aa 27-228. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction