missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP2 Antibody (OTI1C12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70258H
This item is not returnable.
View return policy
Description
BNIP2 Monoclonal specifically detects BNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| BNIP2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| BNIP2 | |
| Full length recombinant protein of human BNIP2 (NP_004321) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1C12 | |
| Western Blot | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 2, BCL2/adenovirus E1B 19kDa interacting protein 2, BCL2/adenovirus E1B 19kD-interacting protein 2, BNIP-2, NIP2 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 663.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction