Learn More
BNIP2 Mouse anti-Human, Clone: OTI1C12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806500
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. Though the specific function is unknown, it interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector.Specifications
| BNIP2 | |
| Monoclonal | |
| Unconjugated | |
| BNIP2 | |
| BNIP-2, NIP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 663 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1C12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q12982 | |
| BNIP2 | |
| Full length human recombit protein of human BNIP2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.